Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030749.2 | NP_109674.2 | 2 | 209 | ||
GenBank mRNA | AJ297884.1 | - | 2 | 150 | ||
AK170415.1 | - | 2 | 162 | |||
BC016119.1 | - | 2 | 186 | 4489217 | ||
BC016466.1 | - | 2 | 172 | 3582798 | ||
BC028488.1 | - | 2 | 146 | |||
CX219879.1 | - | 2 | 209 |