Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfcp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023755.2 | NP_076244.2 | 7 | 765 | ||
GenBank mRNA | AK143960.1 | - | 7 | 764 | ||
BC116663.1 | - | 7 | 760 | 40087602 |