Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031179.2 | NP_112456.2 | 3 | 353 | ||
GenBank mRNA | AB037890.1 | - | 3 | 378 | ||
AK012632.1 | - | 3 | 312 | |||
AK140982.1 | - | 3 | 271 | |||
AK146989.1 | - | 3 | 262 | |||
AK165992.1 | - | 3 | 357 | |||
AK171454.1 | - | 3 | 271 | |||
BC003419.1 | - | 3 | 256 | |||
BC028959.1 | - | 3 | 256 | |||
BC037098.1 | - | 3 | 340 | |||
BC058563.1 | - | 3 | 256 | |||
BC060125.1 | - | 3 | 249 | |||
BC080754.1 | - | 3 | 272 | |||
CK331410.1 | - | 3 | 353 |