Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ucn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031250.5 | NP_112540.2 | 2 | 951 | ||
GenBank mRNA | AK148451.1 | - | 2 | 952 | ||
BC119433.1 | - | 1 | 436 | 40041175 | ||
BC119434.1 | - | 1 | 433 |