Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114385.1 | NP_001107857.1 | 4 | 744 | ||
NM_031258.3 | NP_112548.2 | 4 | 744 | |||
GenBank mRNA | AF296451.1 | - | 4 | 666 | ||
AF305714.1 | - | 3 | 283 | |||
AF321853.1 | - | 3 | 220 | |||
AK142805.1 | - | 4 | 368 | |||
AK164661.1 | - | 4 | 339 | |||
BC050818.1 | - | 3 | 234 | |||
BC066832.1 | - | 4 | 378 | 30102312 |