Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin28a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145833.1 | NP_665832.1 | 4 | 689 | ||
GenBank mRNA | AB221669.1 | - | 4 | 614 | ||
AF521097.1 | - | 4 | 689 | |||
BC068304.1 | - | 4 | 646 | 30432006 |