Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031391.2 | NP_113568.2 | 7 | 751 | ||
NM_175335.3 | NP_780544.1 | 7 | 738 | |||
GenBank mRNA | AF250834.1 | - | 7 | 681 | ||
AK048711.1 | - | 7 | 739 | |||
AK139996.1 | - | 7 | 751 | |||
BC079909.1 | - | 7 | 684 | 30658355 | ||
BC117829.1 | - | 7 | 708 | 40092917 | ||
BC117830.1 | - | 7 | 708 | 40092921 |