Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032002.2 | NP_114391.1 | 3 | 218 | ||
GenBank mRNA | AF083067.2 | - | 2 | 96 | ||
AK041077.1 | - | 3 | 176 | |||
AK080089.1 | - | 3 | 196 | |||
AK080161.1 | - | 3 | 176 | |||
BC034839.1 | - | 3 | 180 | 3372942 |