Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023277.4 | NP_075766.1 | 4, 5 | 454 | ||
GenBank mRNA | AJ300304.1 | - | 4, 5 | 414 | ||
AK003326.1 | - | 4, 5 | 414 | |||
AK008187.1 | - | 4, 5 | 447 | |||
AK013156.1 | - | 4, 5 | 413 | |||
AK032833.1 | - | 4, 5 | 414 | |||
AK160648.1 | - | 4, 5 | 452 | |||
BC024357.1 | - | 4, 5 | 426 | 4973185 |