Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi4k2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145501.2 | NP_663476.1 | 4 | 858 | ||
GenBank mRNA | AK030725.1 | - | 4 | 823 | ||
BC089309.1 | - | 4 | 799 |