Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053107.4 | NP_444337.3 | 2 | 1014 | ||
GenBank mRNA | AK039769.1 | - | 2 | 924 | ||
BC118506.1 | - | 1 | 544 | 40106287 | ||
BC118510.1 | - | 1 | 544 | 40106284 |
Set Membership: |
G-protein coupled receptor |