Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033563.2 | NP_291041.2 | 2 | 1060 | ||
GenBank mRNA | AB221646.1 | - | 2 | 672 | ||
AF327568.1 | - | 2 | 951 | |||
AF338369.1 | - | 2 | 1095 | |||
AK036514.1 | - | 1 | 58 | |||
AK086122.1 | - | 1 | 15 | |||
AK149362.1 | - | 3 | 851 | |||
BC004700.1 | - | 3 | 825 | 3499191 | ||
BC019604.1 | - | 2 | 1060 | 3601534 | ||
BC040794.1 | - | 3 | 825 | 3499191 |