Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnase2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053113.2 | NP_444343.2 | 2 | 414 | ||
GenBank mRNA | BC027557.1 | - | 2 | 414 | 1445853 |