Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159589.1 | NP_001153061.1 | 5 | 1034 | ||
NM_019812.2 | NP_062786.1 | 6 | 1148 | |||
GenBank mRNA | AF214646.1 | - | 6 | 1133 | ||
AK144203.1 | - | 5 | 1037 | |||
AY377984.1 | - | 6 | 1133 | |||
BC152314.1 | - | 6 | 628 | 40111573 |