Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163282.2 | NP_001156754.1 | 1 | 139 | ||
GenBank mRNA | AK162823.1 | - | 1 | 122 | ||
BC094351.1 | - | 1 | 87 | |||
M62301.1 | - | 1 | 127 |
Gene Symbol: Cers1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138647.3 | NP_619588.1 | 1 | 139 | ||
GenBank mRNA | AK053885.1 | - | 1 | 141 | ||
BB665603.1 | - | 1 | 139 | |||
BC132318.1 | - | 1 | 84 | 40130595 |