Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163282.2 | NP_001156754.1 | 5 | 921 | ||
GenBank mRNA | AK162823.1 | - | 5 | 904 | ||
BC094351.1 | - | 5 | 869 | |||
M62301.1 | - | 5 | 909 |
Gene Symbol: Cers1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138647.3 | NP_619588.1 | 5 | 921 | ||
GenBank mRNA | AK053885.1 | - | 5 | 924 | ||
BC132318.1 | - | 5 | 866 | 40130595 |