Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172098.1 | NP_001165569.1 | 8 | 1238 | ||
NM_052973.2 | NP_443205.1 | 10 | 1490 | |||
GenBank mRNA | AF307777.1 | - | 10 | 1474 | ||
AK140447.1 | - | 8 | 1226 | |||
AK165908.1 | - | 8 | 922 | |||
BC138055.1 | - | 8 | 1189 | 8861074 | ||
BC145127.1 | - | 8 | 1189 | 9053675 | ||
EF685152.1 | - | 11 | 1391 | |||
EF685154.1 | - | 3 | 311 |