Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srd5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053188.2 | NP_444418.1 | 2 | 514 | ||
GenBank mRNA | AB049456.1 | - | 2 | 514 | ||
AK138946.1 | - | 2 | 455 | |||
BC125510.1 | - | 2 | 498 | 40130025 |