Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubqln4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033526.2 | NP_277068.1 | 5 | 884 | ||
GenBank mRNA | AB040050.1 | - | 5 | 884 | ||
AK150211.1 | - | 5 | 822 | |||
AK156515.1 | - | 5 | 838 | |||
AK160578.1 | - | 5 | 836 | |||
AK211295.1 | - | 2 | 276 | |||
BC017686.1 | - | 5 | 846 | 4236887 |