Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cadm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053199.3 | NP_444429.1 | 2 | 339 | ||
GenBank mRNA | AF195662.1 | - | 2 | 283 | ||
AK036352.1 | - | 3 | 395 | |||
AK038917.1 | - | 2 | 296 | |||
AK053077.1 | - | 2 | 293 | |||
AY059393.1 | - | 2 | 138 | |||
BC079837.1 | - | 3 | 379 | |||
BC115417.2 | - | 2 | 244 | 40093410 | ||
BC115418.2 | - | 2 | 244 | 40093417 | ||
BY289385.1 | - | 2 | 341 |