Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168256.1 | NP_001161728.1 | 2 | 246 | ||
NM_001168257.1 | NP_001161729.1 | 2 | 246 | |||
NM_144805.2 | NP_659054.2 | 2 | 114 | |||
GenBank mRNA | BC019416.1 | - | 2 | 76 | 5151159 | |
BY104106.1 | - | 2 | 101 | |||
BY279100.1 | - | 2 | 246 | |||
CT010303.1 | - | 1 | 44 | |||
CV561368.1 | - | 2 | 188 | |||
CX564997.1 | - | 2 | 145 |