Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C77080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033189.3 | NP_001028361.1 | 6 | 2616 | ||
NM_001285865.1 | NP_001272794.1 | 6 | 2506 | |||
NM_001285866.1 | NP_001272795.1 | 6 | 2478 | |||
NM_001285867.1 | NP_001272796.1 | 6 | 2367 | |||
GenBank mRNA | AK038020.1 | - | 6 | 2457 | ||
AK129382.1 | - | 6 | 2454 | |||
AK154456.1 | - | 6 | 2368 | |||
AK154932.1 | - | 6 | 2617 |