Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A430005L14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163019.1 | NP_001156491.1 | 2 | 156 | ||
NM_175287.4 | NP_780496.2 | 3 | 270 | |||
GenBank mRNA | AK039812.1 | - | 3 | 271 | ||
BC052148.1 | - | 3 | 270 | 6706797 | ||
BY781135.1 | - | 2 | 128 |