Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133798.3 | NP_598559.2 | 8, 9 | 1973 | ||
GenBank mRNA | AK138497.1 | - | 1249 | |||
AK146901.1 | - | 8, 9 | 1854 | |||
AK147922.1 | - | 8, 9 | 1834 | |||
AK158201.1 | - | 8, 9 | 1864 | |||
AK162206.1 | - | 7, 8 | 1741 | |||
AK164329.1 | - | 8, 9 | 1849 | |||
AK167692.1 | - | 8, 9 | 1974 | |||
BC018508.1 | - | 7, 8 | 1516 | 3156447 |