Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem132a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133804.2 | NP_598565.2 | 4, 5 | 999 | ||
GenBank mRNA | AK032956.1 | - | 4, 5 | 1000 | ||
AK036651.1 | - | 4, 5 | 999 | |||
AK048556.1 | - | 4, 5 | 1000 | |||
AK164646.1 | - | 4, 5 | 1001 | |||
AK173207.1 | - | 4, 5 | 987 | |||
BC006896.1 | - | 4, 5 | 951 | 3599652 |