Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172054.4 | NP_742051.1 | 2 | 137 | ||
GenBank mRNA | AK002893.1 | - | 2 | 142 | ||
AK006170.1 | - | 2 | 192 | |||
AK010709.1 | - | 2 | 136 | |||
AK011424.1 | - | 2 | 81 | |||
AK016756.1 | - | 2 | 124 | |||
AK028525.1 | - | 2 | 134 | |||
AK153857.1 | - | 2 | 138 | |||
AK167045.1 | - | 2 | 101 | |||
BC022947.1 | - | 2 | 118 | 3584012 | ||
BC083077.1 | - | 2 | 134 | 6466275 |