Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133810.3 | NP_598571.2 | 4, 5 | 659 | ||
GenBank mRNA | AK029551.1 | - | 4, 5 | 600 | ||
AK035693.1 | - | 4, 5 | 627 | |||
AK076932.1 | - | 4, 5 | 626 | |||
AK083332.1 | - | 4, 5 | 628 | |||
AK153108.1 | - | 4, 5 | 603 | |||
AK165291.1 | - | 4, 5 | 599 | |||
AY092028.1 | - | 4, 5 | 612 | |||
BC006579.1 | - | 4, 5 | 614 | 3586767 |