Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098528.2 | NP_001091998.1 | 2 | 771 | ||
GenBank mRNA | BC146609.1 | - | 2 | 771 | 40060460 |