Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133838.4 | NP_598599.2 | 5 | 1140 | ||
GenBank mRNA | AB239915.1 | - | 5 | 1051 | ||
AB239916.1 | - | 5 | 1051 | |||
AF173639.1 | - | 5 | 1056 | |||
AK147186.1 | - | 5 | 1092 | |||
AK149760.1 | - | 5 | 1084 | |||
AK159235.1 | - | 5 | 1101 | |||
AK159597.1 | - | 5 | 1100 | |||
AK166101.1 | - | 5 | 1151 | |||
AK169343.1 | - | 5 | 1151 | |||
AK218620.1 | - | 2 | 188 | |||
BC007480.1 | - | 5 | 1180 | 3493227 | ||
BC012272.1 | - | 5 | 1073 | 3485836 |