Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172118.1 | NP_742116.1 | 2 | 123 | ||
GenBank mRNA | AK007972.1 | - | 2 | 133 | ||
AK020258.1 | - | 2 | 124 | |||
AK159880.1 | - | 2 | 168 | |||
BC049974.1 | - | 2 | 158 | 4912899 | ||
BC055439.1 | - | 2 | 147 | 2644903 |