Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153126.2 | NP_694766.1 | 28 | 3158 | ||
GenBank mRNA | AK036294.1 | - | 27 | 3411 | ||
AK040070.1 | - | 27 | 3470 | |||
AK048552.1 | - | 26 | 3340 | |||
AK054029.1 | - | 28 | 3179 | |||
AK129427.1 | - | 1949 | ||||
BC034516.1 | - | 28 | 3158 | 4458024 | ||
BC047436.1 | - | 16 | 1691 |