Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssx2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253768.1 | NP_001240697.1 | 15 | 2226 | ||
NM_001253769.1 | NP_001240698.1 | 14 | 2122 | |||
NM_001253770.1 | NP_001240699.1 | 14 | 1971 | |||
NM_138744.3 | NP_620083.1 | 14 | 1974 | |||
GenBank mRNA | AF532969.1 | - | 13 | 1769 | ||
AK031356.1 | - | 15 | 1950 | |||
AK043865.1 | - | 14 | 2075 | |||
AK049080.1 | - | 14 | 2132 | |||
AK077112.1 | - | 15 | 2011 | |||
AK173064.1 | - | 14 | 1961 | |||
BC021749.1 | - | 14 | 1929 | 4165430 | ||
BC031527.1 | - | 14 | 1937 | 4008624 |