Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Garnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178888.4 | NP_849219.2 | 29 | 2981 | ||
GenBank mRNA | AK034068.1 | - | 29 | 2983 | ||
AK036258.1 | - | 28 | 3279 | |||
AK083673.1 | - | 13 | 1553 | |||
AK133163.1 | - | 30 | 3155 | |||
BC132143.1 | - | 29 | 2924 | 40130420 |