Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026992.3 | NP_081268.1 | 4, 5 | 814 | ||
GenBank mRNA | AK006521.1 | - | 4, 5 | 815 | ||
AK011571.1 | - | 3, 4 | 389 | |||
AY028460.1 | - | 4, 5 | 496 | |||
BC030072.1 | - | 4, 5 | 522 | 3376358 | ||
BC091774.1 | - | 4, 5 | 380 | 5150845 |