Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080968.1 | NP_001074437.1 | 21, 22 | 2269 | ||
NM_133852.2 | NP_598613.2 | 22, 23 | 2350 | |||
GenBank mRNA | AK076370.1 | - | 16, 17 | 1968 | ||
AK220509.1 | - | 22, 23 | 2278 | |||
BC011407.1 | - | 20, 21 | 2070 | 3596079 |