Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271118.1 | NP_001258047.1 | 9 | 2103 | ||
NM_001271119.1 | NP_001258048.1 | 8 | 2088 | |||
NM_001271120.1 | NP_001258049.1 | 9 | 2230 | |||
NM_001271121.1 | NP_001258050.1 | 10 | 1532 | |||
NM_001271122.1 | NP_001258051.1 | 12 | 1698 | |||
NM_001271125.1 | NP_001258054.1 | 14 | 1854 | |||
NM_001271126.1 | NP_001258055.1 | 13 | 1658 | |||
NM_001271127.1 | NP_001258056.1 | 13 | 1712 | |||
NM_001271128.1 | NP_001258057.1 | 13 | 1785 | |||
NM_001271129.1 | NP_001258058.1 | 12 | 1643 | |||
NM_001271130.1 | NP_001258059.1 | 13 | 1728 | |||
NM_031588.2 | NP_113776.2 | 12 | 1634 | |||
GenBank mRNA | AF194438.1 | - | 9 | 2117 | ||
AF194439.1 | - | 8 | 1836 | |||
AF194993.1 | - | 13 | 1728 | |||
AY995221.1 | - | 13 | 1314 | |||
AY995222.1 | - | 10 | 1188 | |||
U02316.1 | - | 13 | 1785 | |||
U02317.1 | - | 13 | 1785 | |||
U02318.1 | - | 12 | 1634 | |||
U02319.1 | - | 12 | 1643 | |||
U02321.1 | - | 12 | 1634 | |||
U02322.1 | - | 13 | 1712 | |||
U02323.1 | - | 12 | 1643 |