Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080781.2 | NP_542959.1 | 19 | 2693 | ||
XM_008759660.1 | XP_008757882.1 | 19 | 2696 | |||
XM_008759661.1 | XP_008757883.1 | 19 | 2783 | |||
GenBank mRNA | BC061882.1 | - | 19 | 2693 | 5599357 | |
X57228.1 | - | 19 | 2624 |