Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053646.1 | NP_446098.1 | 6 | 765 | ||
XM_006231261.2 | XP_006231323.1 | 9 | 1123 | |||
XM_006231263.1 | XP_006231325.1 | 8 | 1701 | |||
XM_006231267.2 | XP_006231329.1 | 8 | 1707 | |||
XM_006231268.2 | XP_006231330.1 | 8 | 1704 | |||
XM_006231270.1 | XP_006231332.1 | 8 | 1704 | |||
XM_008760319.1 | XP_008758541.1 | 11 | 3543 | |||
XM_008760320.1 | XP_008758542.1 | 8 | 984 | |||
GenBank mRNA | AB057433.1 | - | 6 | 765 |