Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053655.3 | NP_446107.2 | 16 | 1971 | ||
XM_006248715.2 | XP_006248777.1 | 18 | 2099 | |||
XM_006248716.2 | XP_006248778.1 | 18 | 2081 | |||
XM_006248717.2 | XP_006248779.1 | 17 | 2065 | |||
XM_006248718.2 | XP_006248780.1 | 17 | 2059 | |||
XM_006248719.2 | XP_006248781.1 | 17 | 2043 | |||
XM_006248720.2 | XP_006248782.1 | 17 | 2020 | |||
XM_006248721.2 | XP_006248783.1 | 16 | 1987 | |||
XM_006248722.2 | XP_006248784.1 | 16 | 1981 | |||
XM_006248723.2 | XP_006248785.1 | 16 | 1969 | |||
XM_006248724.2 | XP_006248786.1 | 15 | 1932 | |||
XM_008768840.1 | XP_008767062.1 | 18 | 2099 | |||
XM_008768841.1 | XP_008767063.1 | 18 | 2081 | |||
XM_008768842.1 | XP_008767064.1 | 17 | 2060 | |||
XM_008768843.1 | XP_008767065.1 | 17 | 2048 | |||
XM_008768844.1 | XP_008767066.1 | 17 | 2043 | |||
XM_008768845.1 | XP_008767067.1 | 17 | 2020 | |||
XM_008768846.1 | XP_008767068.1 | 16 | 2010 | |||
XM_008768847.1 | XP_008767069.1 | 16 | 1987 | |||
GenBank mRNA | AF020211.1 | - | 3 | 333 | ||
AF020212.1 | - | 4 | 411 | |||
AF020213.1 | - | 5 | 444 | |||
BC085843.1 | - | 16 | 1971 | 7135909 |