Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sardh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053664.2 | NP_446116.1 | 21 | 2879 | ||
XM_006233731.2 | XP_006233793.1 | 22 | 2901 | |||
XM_006233732.2 | XP_006233794.1 | 22 | 3337 | |||
XM_006233733.2 | XP_006233795.1 | 22 | 2903 | |||
GenBank mRNA | AF067650.1 | - | 21 | 2914 | ||
FQ218777.1 | - | 21 | 3175 |