Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053675.2 | NP_446127.3 | 3 | 777 | ||
XM_006235636.2 | XP_006235698.1 | 3 | 688 | |||
GenBank mRNA | AF123651.1 | - | 2 | 578 | ||
BC081752.1 | - | 3 | 777 | 7134513 |