Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053702.3 | NP_446154.3 | 4 | 766 | ||
XM_006232266.2 | XP_006232328.1 | 5 | 903 | |||
XM_008760903.1 | XP_008759125.1 | 5 | 898 | |||
GenBank mRNA | AF367448.1 | - | 4 | 665 | ||
AY623027.1 | - | 3 | 740 | |||
AY623028.1 | - | 4 | 1145 | |||
BC127462.1 | - | 5 | 859 | 8373429 | ||
CV114678.1 | - | 1 | 33 | |||
DY551463.1 | - | 4 | 477 |