Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_040669.1 | NP_414541.1 | 11 | 1137 | ||
XM_006231378.2 | XP_006231440.1 | 12 | 1264 | |||
XM_006231379.2 | XP_006231441.1 | 12 | 1390 | |||
XM_006231380.2 | XP_006231442.1 | 12 | 1220 | |||
XM_006231381.2 | XP_006231443.1 | 12 | 1331 | |||
GenBank mRNA | AF333325.1 | - | 11 | 1137 | ||
BC099744.1 | - | 11 | 1167 | 7938691 |