Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kidins220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053795.1 | NP_446247.1 | 20, 21 | 2808 | ||
XM_006239923.2 | XP_006239985.1 | 19, 20 | 3207 | |||
XM_006239924.2 | XP_006239986.1 | 19, 20 | 3204 | |||
XM_006239925.2 | XP_006239987.1 | 20, 21 | 2897 | |||
XM_006239926.2 | XP_006239988.1 | 19, 20 | 3207 | |||
XM_006239927.2 | XP_006239989.1 | 20, 21 | 2894 | |||
XM_006239928.2 | XP_006239990.1 | 20, 21 | 2894 | |||
XM_006239929.2 | XP_006239991.1 | 19, 20 | 3207 | |||
XM_006239931.2 | XP_006239993.1 | 19, 20 | 3207 | |||
XM_006239932.2 | XP_006239994.1 | 19, 20 | 3207 | |||
XM_006239933.2 | XP_006239995.1 | 19, 20 | 3206 | |||
XM_006239934.2 | XP_006239996.1 | 19, 20 | 3206 | |||
XM_006239935.2 | XP_006239997.1 | 19, 20 | 3206 | |||
XM_006239936.2 | XP_006239998.1 | 19, 20 | 3206 | |||
XM_006239937.2 | XP_006239999.1 | 20, 21 | 2897 | |||
GenBank mRNA | AF239045.2 | - | 20, 21 | 2808 | ||
AF313464.1 | - | 19, 20 | 2700 |