Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053819.1 | NP_446271.1 | 5, 6 | 489 | ||
XM_006256608.2 | XP_006256670.1 | 5, 6 | 553 | |||
XM_008773034.1 | XP_008771256.1 | 4, 5 | 463 | |||
GenBank mRNA | AF411319.1 | - | 2, 3 | 239 | ||
AY550026.1 | - | 4, 5 | 478 | |||
BC099821.1 | - | 5, 6 | 487 | 7930800 | ||
FQ219856.1 | - | 5, 6 | 499 | |||
FQ221763.1 | - | 5, 6 | 499 | |||
FQ222271.1 | - | 5, 6 | 499 | |||
FQ222437.1 | - | 5, 6 | 502 | |||
FQ222915.1 | - | 5, 6 | 499 | |||
FQ228400.1 | - | 5, 6 | 498 | |||
FQ230039.1 | - | 5, 6 | 496 | |||
L29512.1 | - | 4, 5 | 367 | |||
L31883.1 | - | 5, 6 | 489 | |||
U06179.1 | - | 4, 5 | 472 |