Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053831.1 | NP_446283.1 | 4 | 1306 | ||
XM_008773386.1 | XP_008771608.1 | 3 | 1049 | |||
GenBank mRNA | L36030.1 | - | 4 | 1306 |