Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053845.1 | NP_446297.1 | 9 | 965 | ||
GenBank mRNA | BC078767.1 | - | 9 | 955 | 7130296 | |
FQ209442.1 | - | 9 | 943 | |||
M97662.1 | - | 9 | 965 |