Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053907.1 | NP_446359.1 | 2 | 382 | ||
GenBank mRNA | AF039852.1 | - | 2 | 332 | ||
BC088122.1 | - | 2 | 380 | 7325326 | ||
FQ226037.1 | - | 2 | 209 | |||
FQ227612.1 | - | 2 | 209 | |||
FQ230283.1 | - | 2 | 209 | |||
FQ230990.1 | - | 2 | 209 | |||
FQ231646.1 | - | 2 | 208 | |||
FQ231744.1 | - | 2 | 208 | |||
FQ231870.1 | - | 2 | 212 | |||
FQ231939.1 | - | 2 | 209 | |||
FQ233228.1 | - | 2 | 265 | |||
FQ234709.1 | - | 2 | 265 | |||
U75689.1 | - | 2 | 382 |