Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053908.1 | NP_446360.1 | 6 | 839 | ||
GenBank mRNA | BC099824.1 | - | 6 | 696 | 7932620 | |
U77038.1 | - | 6 | 839 |