Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054011.1 | NP_446463.2 | 5, 6 | 629 | ||
XM_008770976.1 | XP_008769198.1 | 3, 4 | 2491 | |||
GenBank mRNA | AB027562.1 | - | 5, 6 | 629 | ||
FQ210473.1 | - | 6, 7 | 849 |